CAS RN 64-65-3
CAS RN 64-65-3
IUPAC Name:
4-ethyl-4-methylpiperidine-2,6-dione
Synonyms:
3-ethyl-3-methylglutarimide bemegride (inn) and its salts
Molecular Weight (g/mol):
155.2
Molecular Formula:
C8H13NO2
InChi Key:
ORRZGUBHBVWWOP-UHFFFAOYSA-N
SMILES:
CCC1(C)CC(=O)NC(=O)C1
1
–
3
of
3
results
1
Bemegride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Bemegride, LoGiCal
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
3
Bemegride, MedChemExpress
MedChemExpress Bemegride (3-Ethyl-3-methylglutarimide) is a central nervous system stimulant and antidote for barbiturate poisoning.