accessibility menu, dialog, popup

UserName

CAS RN 64051-79-2

HClOHHN

CAS RN 64051-79-2

IUPAC Name: hydrogen piperidin-3-ol chloride
Synonyms: hydrogen piperidin-3-ol chloride
Molecular Weight (g/mol): 137.61
Molecular Formula: C5H12ClNO
InChi Key: VLECDMDGMKPUSK-UHFFFAOYNA-N
SMILES: [H+].[Cl-].OC1CCCNC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)