missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Hydroxypiperidine hydrochloride, 99%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 196.6°C to 198.7°C |
| Color | White |
| Elemental Analysis | 42.97 to 44.28% C, 9.90 to 10.33% N |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Quantity | 25 g |
| Formula Weight | 137.61 |
| Percent Purity | 99% |
| Show More |
Safety and Handling
Product Identifier
- 3-Hydroxypiperidine hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction