accessibility menu, dialog, popup

UserName

CAS RN 64109-12-2

H2OPdO

CAS RN 64109-12-2

IUPAC Name: oxopalladium hydrate
Synonyms: oxopalladium hydrate
Molecular Weight (g/mol): 140.43
Molecular Formula: H2O2Pd
InChi Key: CJCUJYURPLYQOL-UHFFFAOYSA-N
SMILES: O.O=[Pd]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results