accessibility menu, dialog, popup

UserName

CAS RN 64628-73-5

NH2OFFFCl

CAS RN 64628-73-5

IUPAC Name: 3-chloro-4-(trifluoromethoxy)aniline
Synonyms: 3-chloro-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 211.57
Molecular Formula: C7H5ClF3NO
InChi Key: ZPKUUNGPBSRPRM-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC(F)(F)F)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results