CAS RN 64987-03-7
IUPAC Name:
ethyl 2-(2-formamido-1,3-thiazol-4-yl)-2-oxoacetate
Synonyms:
ethyl 2-(2-formamido-1,3-thiazol-4-yl)-2-oxoacetate
Molecular Weight (g/mol):
228.22
Molecular Formula:
C8H8N2O4S
InChi Key:
NTACMHVXGGGRQU-UHFFFAOYSA-N
SMILES:
CCOC(=O)C(=O)C1=CSC(NC=O)=N1
1
–
3
of
3
results
1
Ethyl 2-(2-Formamidothiazol-4-yl)-2-oxoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Ethyl 2-(2-Formamidothiazol-4-yl)-2-oxoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Ethyl 2-(2-Formamidothiazol-4-yl)-2-oxoacetate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.