CAS RN 65813-55-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-Oxo Ibuprofen, TRC
CAS: 65813-55-0 Molecular Formula: C13 H16 O3 Molecular Weight (g/mol): 220.26 Synonym: (2RS)-2-[4-(2-Methyl-propanoyl)phenyl]propanoic Acid,Ibuprofen Imp. J (EP) IUPAC Name: 2-[4-(2-methylpropanoyl)phenyl]propanoic acid SMILES: CC(C)C(=O)c1ccc(cc1)C(C)C(=O)O
2
(2RS)-2-[4-(2-Methyl-propanoyl)phenyl]propanoic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.