CAS RN 66648-50-8
IUPAC Name:
ethyl (2E)-3-(3,4-dihydroxyphenyl)prop-2-enoate
Synonyms:
ethyl caffeate ethyl cinnamate,3,4-dihydroxy
Molecular Weight (g/mol):
208.21
Molecular Formula:
C11H12O4
InChi Key:
WDKYDMULARNCIS-GQCTYLIASA-N
SMILES:
CCOC(=O)\C=C\C1=CC=C(O)C(O)=C1
1
–
1
of
1
results
1
Ethyl 3,4-dihydroxycinnamate, 97%
CAS: 66648-50-8 Molecular Formula: C11H12O4 Molecular Weight (g/mol): 208.213 MDL Number: MFCD00045754 InChI Key: WDKYDMULARNCIS-GQCTYLIASA-N Synonym: ethyl caffeate,ethyl 3-3,4-dihydroxyphenyl acrylate,ethyl 3,4-dihydroxycinnamate,caffeic acid ethyl ester,ethyl trans-caffeate,unii-76gbb1ju5y,e-ethyl 3-3,4-dihydroxyphenyl acrylate,3,4-dihydroxycinnamic acid ethyl ester,76gbb1ju5y,chembl17347 PubChem CID: 5317238 ChEBI: CHEBI:69656 IUPAC Name: ethyl (E)-3-(3,4-dihydroxyphenyl)prop-2-enoate SMILES: CCOC(=O)C=CC1=CC(=C(C=C1)O)O