accessibility menu, dialog, popup

UserName

H3COFFFBr

CAS RN 685126-88-9

IUPAC Name: 2-(bromomethyl)-1-methoxy-4-(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-1-methoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 269.06
Molecular Formula: C9H8BrF3O
InChi Key: ZGKOXCPOSPRGFQ-UHFFFAOYSA-N
SMILES: COC1=C(CBr)C=C(C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results