CAS RN 68573-24-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
N-(p-Coumaroyl) Serotonin, TRC
CAS: 68573-24-0 Molecular Formula: C19 H18 N2 O3 Molecular Weight (g/mol): 322.36 Synonym: 2-Propenamide, N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]-3-(4-hydroxyphenyl)-,N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]-3-(4-hydroxyphenyl)-2-propenamide,N-(p-Coumaroyl) serotonin,N-(p-Coumaroyl)serotonin,NSC 369503 IUPAC Name: (E)-N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]-3-(4-hydroxyphenyl)prop-2-enamide SMILES: Oc1ccc(\C=C\C(=O)NCCc2c[nH]c3ccc(O)cc23)cc1