missing translation for 'onlineSavingsMsg'
Learn More

N-(p-Coumaroyl) Serotonin, TRC

Product Code. 30368649
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
10 mg
Unit Size:
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30368649 10 mg -
30350671 5 mg 5mg
30353423 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30368649 Supplier Toronto Research Chemicals Supplier No. TRCC75630010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68573-24-0
Molecular Formula C19 H18 N2 O3
Molecular Weight (g/mol) 322.36
Synonym 2-Propenamide, N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]-3-(4-hydroxyphenyl)-, N-[2-(5-Hydroxy-1H-indol-3-yl)ethyl]-3-(4-hydroxyphenyl)-2-propenamide, N-(p-Coumaroyl) serotonin, N-(p-Coumaroyl)serotonin, NSC 369503
IUPAC Name (E)-N-[2-(5-hydroxy-1H-indol-3-yl)ethyl]-3-(4-hydroxyphenyl)prop-2-enamide
SMILES Oc1ccc(\C=C\C(=O)NCCc2c[nH]c3ccc(O)cc23)cc1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C19H18N2O3/c22-15-4-1-13(2-5-15)3-8-19(24)20-10-9-14-12-21-18-7-6-16(23)11-17(14)18/h1-8,11-12,21-23H,9-10H2,(H,20,24)/b8-3+
Formula Weight 322.1317
Chemical Name or Material N-(p-Coumaroyl) Serotonin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.