CAS RN 690955-62-5
CAS RN 690955-62-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Olpadronic Acid Ammonium, TRC
CAS: 690955-62-5 Molecular Formula: C5 H15 N O7 P2 . x(NH3) Molecular Weight (g/mol): 263.121703 Synonym: P,P'-[3-(Dimethylamino)-1-hydroxypropylidene]bisphosphonic Acid Ammonium Salt,3-(Dimethylamino)-1-hydroxypropylidene-1,1-bisphosphonic Acid Ammonium Salt,3-Dimethylamino-1-hydroxypropylidenebiphosphonic Acid Ammonium Salt; SMILES: OC(P(O)(O)=O)(P(O)(O)=O)CCN(C)C.N