Learn More
Olpadronic Acid Ammonium, TRC
Product Code. 30010506
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010506 1 g 1g
30003692 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30010506 Supplier Toronto Research Chemicals Supplier No. TRCO5753001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 690955-62-5
Molecular Formula C5 H15 N O7 P2 . x(NH3)
Molecular Weight (g/mol) 263.121703
Synonym P,P'-[3-(Dimethylamino)-1-hydroxypropylidene]bisphosphonic Acid Ammonium Salt, 3-(Dimethylamino)-1-hydroxypropylidene-1,1-bisphosphonic Acid Ammonium Salt, 3-Dimethylamino-1-hydroxypropylidenebiphosphonic Acid Ammonium Salt;
SMILES OC(P(O)(O)=O)(P(O)(O)=O)CCN(C)C.N
Specifications
Quantity 1 g
InChI Formula InChI=1S/C5H15NO7P2.H3N/c1-6(2)4-3-5(7,14(8,9)10)15(11,12)13;/h7H,3-4H2,1-2H3,(H2,8,9,10)(H2,11,12,13);1H3
Chemical Name or Material Olpadronic Acid Ammonium Salt