CAS RN 695-98-7
CAS RN 695-98-7
IUPAC Name:
2,3,5-trimethylpyridine
Synonyms:
pyridine, 2,3,5-trimethyl-
Molecular Weight (g/mol):
121.18
Molecular Formula:
C8H11N
InChi Key:
GFYHSKONPJXCDE-UHFFFAOYSA-N
SMILES:
CC1=CN=C(C)C(C)=C1
1
–
2
of
2
results
2
2,3,5-Collidine, 99%
CAS: 695-98-7 Molecular Formula: C8H11N Molecular Weight (g/mol): 121.183 MDL Number: MFCD00126840 InChI Key: GFYHSKONPJXCDE-UHFFFAOYSA-N Synonym: 2,3,5-collidine,pyridine, 2,3,5-trimethyl,2,3,5-trimethyl pyridine,pubchem15901,acmc-1aul1,2,3,5-trimethyl-pyridin,2,3,5-trimethyl-pyridine,ksc490s0f PubChem CID: 12759 IUPAC Name: 2,3,5-trimethylpyridine SMILES: CC1=CC(=C(N=C1)C)C