Learn More
2,3,5-Collidine, 99%
Product Code. 11475547
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 11475547
Brand: Thermo Scientific Alfa Aesar B23537.22
Request Bulk or Custom Format
CAS: 695-98-7 | C8H11N | 121.183 g/mol
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 695-98-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00126840
InChI Key GFYHSKONPJXCDE-UHFFFAOYSA-N
Synonym 2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f
PubChem CID 12759
IUPAC Name 2,3,5-trimethylpyridine
SMILES CC1=CC(=C(N=C1)C)C
Specifications
Density 0.935
Boiling Point 182°C to 184°C
Flash Point 74°C (165°F)
Odor Unpleasant
Refractive Index 1.5075
Quantity 100 g
UN Number UN2810
Beilstein 108832
Solubility Information Slightly soluble in water.
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,3,5-Collidine