accessibility menu, dialog, popup

UserName

CAS RN 695-99-8

ClOO

CAS RN 695-99-8

IUPAC Name: 2-chlorocyclohexa-2,5-diene-1,4-dione
Synonyms: 2-chlorocyclohexa-2,5-diene-1,4-dione
Molecular Weight (g/mol): 142.54
Molecular Formula: C6H3ClO2
InChi Key: WOGWYSWDBYCVDY-UHFFFAOYSA-N
SMILES: ClC1=CC(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)