accessibility menu, dialog, popup

UserName

CAS RN 6968-35-0

OHO

CAS RN 6968-35-0

IUPAC Name: 7-hydroxy-2,3-dihydro-1H-inden-1-one
Synonyms: 7-hydroxy-2,3-dihydroinden-1-one
Molecular Weight (g/mol): 148.16
Molecular Formula: C9H8O2
InChi Key: HFMZPBSZKCDKOR-UHFFFAOYSA-N
SMILES: OC1=CC=CC2=C1C(=O)CC2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)