CAS RN 705946-27-6
CAS RN 705946-27-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tyrosine kinase-IN-1, MedChemExpress
MedChemExpress Tyrosine kinase-IN-1 is a multi-targeted tyrosine kinase inhibitor with IC50s of 4, 20, 4, 2 nM for KDR, Flt-1, FGFR1 and PDGFRα, respectively.