accessibility menu, dialog, popup

UserName

CAS RN 7114-03-6

ClClClClZnH3CNH3CCH3CNCH3H3CNCH3CH3

CAS RN 7114-03-6

IUPAC Name: zinc(2+) bis[4-(dimethylamino)phenyl][4-(ethyldimethylazaniumyl)phenyl]methylium tetrachloride
Synonyms: zinc(2+) bis[4-(dimethylamino)phenyl][4-(ethyldimethylammonio)phenyl]methylium tetrachloride
Molecular Weight (g/mol): 608.78
Molecular Formula: C27H35Cl4N3Zn
InChi Key: ZKOGXCSOLMZRAZ-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Zn++].CC[N+](C)(C)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(C)C)C1=CC=C(C=C1)N(C)C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (3)
  • (1)
Color 
  • (2)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)