accessibility menu, dialog, popup

UserName

CAS RN 71432-55-8

H3CCH3NHNCH3H3COCH3H3CCH3(E/Z)

CAS RN 71432-55-8

IUPAC Name: N,N'-bis(propan-2-yl)(tert-butoxy)methanimidamide
Synonyms: N,N'-diisopropyltert-butoxymethanimidamide
Molecular Weight (g/mol): 200.33
Molecular Formula: C11H24N2O
InChi Key: FESDUDPSRMWIDL-UHFFFAOYSA-N
SMILES: CC(C)NC(OC(C)(C)C)=NC(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results