accessibility menu, dialog, popup

UserName

CH3OOH3CNOCH3H3C

CAS RN 71626-11-4

IUPAC Name: methyl 2-[N-(2,6-dimethylphenyl)-2-phenylacetamido]propanoate
Synonyms: galben epa pesticide chemical code 127001 benalaxyl caswell no. 471AC
Molecular Weight (g/mol): 325.41
Molecular Formula: C20H23NO3
InChi Key: CJPQIRJHIZUAQP-UHFFFAOYNA-N
SMILES: COC(=O)C(C)N(C(=O)CC1=CC=CC=C1)C1=C(C)C=CC=C1C
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
Concentration 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results