CAS RN 71888-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
a-D-Mannose-1-phosphate Dipotassium Salt, TRC
CAS: 71888-67-0 Molecular Formula: C6H11K2O9P Molecular Weight (g/mol): 336.32 IUPAC Name: dipotassium;[(2R,3S,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl] phosphate SMILES: C([C@@H]1[C@H]([C@@H]([C@@H]([C@H](O1)OP(=O)([O-])[O-])O)O)O)O.[K+].[K+]