accessibility menu, dialog, popup

UserName

H2NON

CAS RN 7251-52-7

IUPAC Name: 2-phenyl-2-(pyridin-2-yl)acetamide
Synonyms: α-(2-pyridyl)phenylacetamide
Molecular Weight (g/mol): 212.25
Molecular Formula: C13H12N2O
InChi Key: PYAPITOPBTXXNJ-UHFFFAOYNA-N
SMILES: NC(=O)C(C1=CC=CC=C1)C1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results