accessibility menu, dialog, popup

UserName

CAS RN 7287-82-3

H3COHH3C

CAS RN 7287-82-3

IUPAC Name: 1-(2-methylphenyl)ethan-1-ol
Synonyms: 1-(2-methylphenyl)ethanol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: SDCBYRLJYGORNK-UHFFFAOYNA-N
SMILES: CC(O)C1=CC=CC=C1C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results