accessibility menu, dialog, popup

UserName

CAS RN 73482-96-9

H3COOH3COOH3COOH3COORhRh

CAS RN 73482-96-9

IUPAC Name: bis(λ²-rhodium(2+)) tetraoctanoate
Synonyms: bis(λ²-rhodium(2+)) tetraoctanoate
Molecular Weight (g/mol): 778.64
Molecular Formula: C32H60O8Rh2
InChi Key: FZXFNYFVNKTQSX-UHFFFAOYSA-J
SMILES: [Rh++].[Rh++].CCCCCCCC([O-])=O.CCCCCCCC([O-])=O.CCCCCCCC([O-])=O.CCCCCCCC([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results