accessibility menu, dialog, popup

UserName

CAS RN 736989-98-3

BOHHOOH

CAS RN 736989-98-3

IUPAC Name: [3-(3-hydroxypropyl)phenyl]boronic acid
Synonyms: 3-(3-hydroxypropyl)phenylboronic acid
Molecular Weight (g/mol): 180.01
Molecular Formula: C9H13BO3
InChi Key: OTCBPHAZRFXGKH-UHFFFAOYSA-N
SMILES: OCCCC1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results