accessibility menu, dialog, popup

UserName

CAS RN 7385-10-6

CH3SiH3CCH3

CAS RN 7385-10-6

IUPAC Name: (9H-fluoren-9-yl)trimethylsilane
Synonyms: 9H-fluoren-9-yltrimethylsilane
Molecular Weight (g/mol): 238.41
Molecular Formula: C16H18Si
InChi Key: YFPRGLHBYSNHMW-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C1C2=CC=CC=C2C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results