accessibility menu, dialog, popup

UserName

CAS RN 7451-98-1

CH3OOHHO

CAS RN 7451-98-1

IUPAC Name: 1-(3,4-dihydroxyphenyl)propan-1-one
Synonyms: 1-(3,4-dihydroxyphenyl)propan-1-one
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: HNWIHBDMOYWCGX-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC=C(O)C(O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results