CAS RN 74974-54-2
CAS RN 74974-54-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-1,1,1-trimethoxyethane, TRC
CAS: 74974-54-2 Molecular Formula: C5H11ClO3 Molecular Weight (g/mol): 154.59 Synonym: 1-Chloro-2,2,2-trimethoxyethane,Trimethyl (Chloromethyl)orthoformate,Trimethyl Orthochloroacetate IUPAC Name: 2-chloro-1,1,1-trimethoxyethane SMILES: ClCC(OC)(OC)OC