accessibility menu, dialog, popup

UserName

CAS RN 75422-66-1

CH3SiH3CCH2Cl

CAS RN 75422-66-1

IUPAC Name: (chloromethyl)dimethyl(prop-2-en-1-yl)silane
Synonyms: (chloromethyl)dimethylprop-2-en-1-ylsilane
Molecular Weight (g/mol): 148.71
Molecular Formula: C6H13ClSi
InChi Key: UHHLOVCFFWGSMO-UHFFFAOYSA-N
SMILES: C[Si](C)(CCl)CC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results