accessibility menu, dialog, popup

UserName

HOOFFCl

CAS RN 76-04-0

IUPAC Name: 2-chloro-2,2-difluoroacetic acid
Synonyms: acetic acid, chlorodifluoro-
Molecular Weight (g/mol): 130.47
Molecular Formula: C2HClF2O2
InChi Key: OAWAZQITIZDJRB-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results