accessibility menu, dialog, popup

UserName

CAS RN 761459-04-5

CH3OOClFF

CAS RN 761459-04-5

IUPAC Name: 4-ethoxy-2,3-difluorobenzoyl chloride
Synonyms: 4-ethoxy-2,3-difluorobenzoyl chloride
Molecular Weight (g/mol): 220.6
Molecular Formula: C9H7ClF2O2
InChi Key: BTIZLROTPPAIEJ-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C(F)=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results