missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxy-2,3-difluorobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 15434257
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15434257 5 g 5g
15424257 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15434257

Brand: Thermo Scientific Alfa Aesar H31648.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 761459-04-5
Molecular Formula C9H7ClF2O2
Molecular Weight (g/mol) 220.6
MDL Number MFCD09258701
InChI Key BTIZLROTPPAIEJ-UHFFFAOYSA-N
Synonym 4-ethoxy-2,3-difluorobenzoyl chloride, jrd
PubChem CID 19763022
IUPAC Name 4-ethoxy-2,3-difluorobenzoyl chloride
SMILES CCOC1=C(C(=C(C=C1)C(=O)Cl)F)F

Specifications

Melting Point 49°C to 51°C
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Formula Weight 220.6
Percent Purity 97%
Chemical Name or Material 4-Ethoxy-2,3-difluorobenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.