CAS RN 7622-29-9
IUPAC Name:
hydrogen 2-amino-6-(methylamino)hexanoic acid chloride
Synonyms:
hydrogen 2-amino-6-(methylamino)hexanoic acid chloride
Molecular Weight (g/mol):
196.68
Molecular Formula:
C7H17ClN2O2
InChi Key:
AQELUQTVJOFFBN-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].CNCCCCC(N)C(O)=O
1
–
3
of
3
results
1
Epsilon-n-methyl-l-lysine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Epsilon-n-methyl-l-lysine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Epsilon-n-methyl-l-lysine Hydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.