CAS RN 7643-08-5
CAS RN 7643-08-5
IUPAC Name:
2-(methylsulfanyl)-10H-phenothiazine
Synonyms:
2-(methylsulfanyl)-10H-phenothiazine
Molecular Weight (g/mol):
245.36
Molecular Formula:
C13H11NS2
InChi Key:
OBVKBOLDEFIQDP-UHFFFAOYSA-N
SMILES:
CSC1=CC=C2SC3=CC=CC=C3NC2=C1