CAS RN 76855-69-1
CAS RN 76855-69-1
IUPAC Name:
3-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-4-oxoazetidin-2-yl acetate
Synonyms:
3-{1-[(tert-butyldimethylsilyl)oxy]ethyl}-4-oxoazetidin-2-yl acetate
Molecular Weight (g/mol):
287.43
Molecular Formula:
C13H25NO4Si
InChi Key:
GWHDKFODLYVMQG-UHFFFAOYNA-N
SMILES:
CC(O[Si](C)(C)C(C)(C)C)C1C(NC1=O)OC(C)=O
1
–
2
of
2
results
1
(3R,4R)-4-Acetoxy-3-[(R)-1-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
(3R,4R)-4-Acetoxy-3-[(R)-1-(tert-butyldimethylsilyloxy)ethyl]-2-azetidinone, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.