accessibility menu, dialog, popup

UserName

H3C

CAS RN 770-69-4

IUPAC Name: 1-ethyladamantane
Synonyms: 1-ethyladamantane
Molecular Weight (g/mol): 164.29
Molecular Formula: C12H20
InChi Key: LXTHCCWEYOKFSR-UHFFFAOYSA-N
SMILES: CCC12CC3CC(CC(C3)C1)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results