CAS RN 77095-51-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Benzofuran-6-carboxylic Acid, TRC
CAS: 77095-51-3 Molecular Formula: C9H6O3 Molecular Weight (g/mol): 162.14 Synonym: 6-Benzofurancarboxylic Acid; IUPAC Name: 1-benzofuran-6-carboxylic acid SMILES: OC(=O)c1ccc2ccoc2c1