missing translation for 'onlineSavingsMsg'
Learn More

Benzofuran-6-carboxylic Acid, TRC

Product Code. 30137094
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137094 1 g 1g
30012110 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30137094 Supplier Toronto Research Chemicals Supplier No. TRCB2854201G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77095-51-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
Synonym 6-Benzofurancarboxylic Acid;
IUPAC Name 1-benzofuran-6-carboxylic acid
SMILES OC(=O)c1ccc2ccoc2c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H6O3/c10-9(11)7-2-1-6-3-4-12-8(6)5-7/h1-5H,(H,10,11)
Formula Weight 162.0317
Chemical Name or Material Benzofuran-6-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.