accessibility menu, dialog, popup

UserName

CAS RN 771-56-2

CH3FFFFF

CAS RN 771-56-2

IUPAC Name: 1,2,3,4,5-pentafluoro-6-methylbenzene
Synonyms: N-methylpentafluorobenzene
Molecular Weight (g/mol): 182.09
Molecular Formula: C7H3F5
InChi Key: SXPRVMIZFRCAGC-UHFFFAOYSA-N
SMILES: CC1=C(F)C(F)=C(F)C(F)=C1F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results