missing translation for 'onlineSavingsMsg'
Learn More

2,3,4,5,6-Pentafluorotoluene, 99%

Product Code. 11499387
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 11499387

Brand: Thermo Scientific Alfa Aesar B21419.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 771-56-2 | C7H3F5 | 182.093 g/mol

2,3,4,5,6-Pentafluorotoluene, can be used as an organic fluorinated building blocks for chemical synthesis. It is also used as an pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,4,5,6-Pentafluorotoluene, can be used as an organic fluorinated building blocks for chemical synthesis. It is also used as an pharmaceutical intermediate.

Solubility
It is soluble in most common organic solvents and insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-56-2
Molecular Formula C7H3F5
Molecular Weight (g/mol) 182.093
MDL Number MFCD00000298
InChI Key SXPRVMIZFRCAGC-UHFFFAOYSA-N
Synonym 2,3,4,5,6-pentafluorotoluene, pentafluorotoluene, methyl pentafluorobenzene, pentafluoromethylbenzene, benzene, pentafluoromethyl, methylpentafluorobenzene, benzene, 1,2,3,4,5-pentafluoro-6-methyl, toluene, 2,3,4,5,6-pentafluoro, 2,3,4,5,6-pentafluoro-toluene, n-methylpentafluorobenzene
PubChem CID 69869
IUPAC Name 1,2,3,4,5-pentafluoro-6-methylbenzene
SMILES CC1=C(C(=C(C(=C1F)F)F)F)F

Specifications

Melting Point -30°C
Density 1.439
Boiling Point 116°C to 117°C
Flash Point 34°C (93°F)
Refractive Index 1.403
Quantity 50 g
UN Number UN1993
Beilstein 1911885
Solubility Information It is soluble in most common organic solvents and insoluble in water.
Formula Weight 182.09
Percent Purity 99%
Chemical Name or Material 2,3,4,5,6-Pentafluorotoluene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.