accessibility menu, dialog, popup

UserName

CAS RN 771574-00-6

CH3FNH2

CAS RN 771574-00-6

IUPAC Name: 1-(4-fluoro-2-methylphenyl)methanamine
Synonyms: 1-(4-fluoro-2-methylphenyl)methanamine
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: KMDTUDLZKZTPCP-UHFFFAOYSA-N
SMILES: CC1=C(CN)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results