accessibility menu, dialog, popup

UserName

FO

CAS RN 772-31-6

IUPAC Name: cyclopropyl(4-fluorophenyl)methanone
Synonyms: cyclopropyl(4-fluorophenyl)methanone
Molecular Weight (g/mol): 164.18
Molecular Formula: C10H9FO
InChi Key: MHKHJIJXMVHRAJ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)C(=O)C1CC1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)