accessibility menu, dialog, popup

UserName

KOIOO

CAS RN 7758-05-6

IUPAC Name: potassium iodate
Synonyms: potassium iodate
Molecular Weight (g/mol): 214
Molecular Formula: IKO3
InChi Key: JLKDVMWYMMLWTI-UHFFFAOYSA-M
SMILES: [K+].[O-][I](=O)=O
Molecular Weight (g/mol) 
  • (9)
  • (4)
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (6)
Grade 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (6)
  • (1)
  • (3)
  • (9)
  • (4)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (6)
  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)