CAS RN 77616-97-8
CAS RN 77616-97-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
3-Amino-6-bromo-4-(pyridin-2-yl)quinolin-2(1H)-one, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-Amino-6-bromo-4-(2-pyridinyl)-2(1H)-quinolinone(>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
3-Amino-6-bromo-4-(2-pyridinyl)-2(1H)-quinolinone(>90%), TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.