accessibility menu, dialog, popup

UserName

FFMg

CAS RN 7783-40-6

IUPAC Name: magnesium(2+) difluoride
Synonyms: magnesium fluoride
Molecular Weight (g/mol): 62.3
Molecular Formula: F2Mg
InChi Key: ORUIBWPALBXDOA-UHFFFAOYSA-L
SMILES: [F-].[F-].[Mg++]
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
Physical Form 
  • (2)
  • (6)
  • (2)
Percent Purity 
  • (4)
  • (3)
Grade 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (12)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (6)
  • (2)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)