accessibility menu, dialog, popup

UserName

NaOBrOO

CAS RN 7789-38-0

IUPAC Name: sodium bromate
Synonyms: sodium bromate
Molecular Weight (g/mol): 150.89
Molecular Formula: BrNaO3
InChi Key: XUXNAKZDHHEHPC-UHFFFAOYSA-M
SMILES: [Na+].[O-][Br](=O)=O
Molecular Weight (g/mol) 
  • (3)
  • (7)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (6)
  • (3)
Physical Form 
  • (1)
  • (3)
  • (3)
  • (1)
Grade 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (3)