accessibility menu, dialog, popup

UserName

H3CSClOO

CAS RN 7795-95-1

IUPAC Name: octane-1-sulfonyl chloride
Synonyms: octane-1-sulfonyl chloride
Molecular Weight (g/mol): 212.73
Molecular Formula: C8H17ClO2S
InChi Key: WIVNTNLDTMNDNO-UHFFFAOYSA-N
SMILES: CCCCCCCCS(Cl)(=O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)