missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11492787
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492787 50 g 50g
11482787 10 g 10g
11402797 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492787 Supplier Thermo Scientific Alfa Aesar Supplier No. A15440.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7795-95-1
Molecular Formula C8H17ClO2S
Molecular Weight (g/mol) 212.732
MDL Number MFCD00014729
InChI Key WIVNTNLDTMNDNO-UHFFFAOYSA-N
Synonym 1-octanesulfonyl chloride, n-octylsulfonyl chloride, octanesulfonyl chloride, unii-3sy5pr1ee7, 3sy5pr1ee7, 1-octanesulfonylchloride, 1-octanesulphonyl chloride, chlorooctylsulfone, octane sulfonyl chloride, acmc-209pcx
PubChem CID 82261
IUPAC Name octane-1-sulfonyl chloride
SMILES CCCCCCCCS(=O)(=O)Cl

Specifications

Melting Point 14°C
Density 1.089
Boiling Point 75°C (0.03 mmHg)
Flash Point >110°C (230°F)
Linear Formula CH3(CH2)7SO2Cl
Refractive Index 1.46
Quantity 50 g
UN Number UN3265
Beilstein 1763921
Sensitivity Moisture sensitive
Formula Weight 212.74
Percent Purity 97%
Chemical Name or Material 1-Octanesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Sekretesspolicy.