CAS RN 78210-78-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Methyl-2,3'-bipyridine, TRC
CAS: 78210-78-3 Molecular Formula: C11H10N2 Molecular Weight (g/mol): 170.21 Synonym: 6-Methyl-2,3'-bipyridine,2-Methyl-6-(pyridin-3-yl)pyridine,2-Methyl-6-pyridin-3-ylpyridine SMILES: Cc1cccc(n1)c2cccnc2